Learn More
Cygnus Technologies Sf9 2G HCP ELISA Kit
Supplier:  Cygnus Technologies F840
Encompass
This kit is intended for use in determining the presence of host cell protein (HCP) impurities in products manufactured by recombinant expression in Sf9 insect host cells.
Catalog No. NC1245033