Learn More
New England Biolabs, Inc. SfaNI – 300 units
SDP
Catalog No. 50812083
Encompass
A restriction endonuclease that recognizes the sequence GCATCNNNNN^NNNN_.
Catalog No. 50-812-083 Supplier New England Biolabs, Inc. Supplier No. R0172S
A restriction endonuclease that recognizes the sequence GCATCNNNNN^NNNN_.