Learn More
Biotage Sfär Bio C18 Duo 300 Å 20µm 10g
SDP
Catalog No. NC1606470
Encompass
High performance flash chromatography is a very efficient technique to dramatically clean-up synthetic peptides. Biotage Sfär Bio flash columns were developed with a small particle size (20 μm) and large pore size (300 Å) to provide increased resolution and the effective separation of complex peptide mixtures. Depending on the quality of the peptides synthesized, flash chromatography can be used either as the sole method of purification or as a front end clean-up prior to RP-HPLC.These 10g Biotage Sfär Bio flash C18 columns come in a pack of 2 columns. They allow more peptide to be processed in a single injection and a reduction in solvent usage and waste. Biotage Sfär Bio offers close to prep-HPLC performance, in a flash format at a fraction of the cost. Biotage Sfär columns are available as C18 and C4 media - C4 is slightly more polar than C18, and therefore the selectivity of C4 is increased for smaller and more polar molecules
Catalog No. NC1606470 Supplier Biotage Supplier No. FSBD04110010
High performance flash chromatography is a very efficient technique to dramatically clean-up synthetic peptides. Biotage Sfär Bio flash columns were developed with a small particle size (20 μm) and large pore size (300 Å) to provide increased resolution and the effective separation of complex peptide mixtures. Depending on the quality of the peptides synthesized, flash chromatography can be used either as the sole method of purification or as a front end clean-up prior to RP-HPLC.These 10g Biotage Sfär Bio flash C18 columns come in a pack of 2 columns. They allow more peptide to be processed in a single injection and a reduction in solvent usage and waste. Biotage Sfär Bio offers close to prep-HPLC performance, in a flash format at a fraction of the cost. Biotage Sfär columns are available as C18 and C4 media - C4 is slightly more polar than C18, and therefore the selectivity of C4 is increased for smaller and more polar molecules