Learn More
Biotage Sfär C18 Duo 100 Å 30µm 400g
SDP
Catalog No. NC1606525
Encompass
Biotage Sfär C18 D is the column of choice when you want to increase efficiency and speed in your reverse phase separations. These high capacity columns make it possible to routinely load three times more sample than preparative HPLC. This means that a smaller column will do the job in a single injection and use less solvent along the way.Cost and time are reduced while performance is increased, thanks to spherical 30 µm well-packed silica particles and the new design of the Sfär columns that makes them even more resistant to high pressure (as high as 17 bar or 247 psi)) often generated by high performance C18 media. Also when running reverse phase, thanks to the Duo capability, you have the flexibility to choose how to load your samples, and choose from one of six different ways or use a Biotage Samplet as a guard column to prolong column life.These 400g Biotage Sfar C18 Duo columns come packaged 1 column per box.
Catalog No. NC1606525 Supplier Biotage Supplier No. FSUD04010400
Biotage Sfär C18 D is the column of choice when you want to increase efficiency and speed in your reverse phase separations. These high capacity columns make it possible to routinely load three times more sample than preparative HPLC. This means that a smaller column will do the job in a single injection and use less solvent along the way.Cost and time are reduced while performance is increased, thanks to spherical 30 µm well-packed silica particles and the new design of the Sfär columns that makes them even more resistant to high pressure (as high as 17 bar or 247 psi)) often generated by high performance C18 media. Also when running reverse phase, thanks to the Duo capability, you have the flexibility to choose how to load your samples, and choose from one of six different ways or use a Biotage Samplet as a guard column to prolong column life.These 400g Biotage Sfar C18 Duo columns come packaged 1 column per box.