Learn More
Biotage Sfär C18 Samplet 50 to 120g Column
SDP
Catalog No. NC1607503
Encompass
Creating dried, adsorbed samples will improve the separation performance on any cartridge. Biotage Samplet are designed for quick and convenient loading of concentrated samples.After the sample has been applied to the Samplet, the solvent is allowed to evaporate. The Samplet is then inserted into the Biotage Sfär Duo columns and you are ready to go.Samplets also are used as guard columns for Biotage Sfar C18, Biotage Sfar Bio C18 and Biotage Sfar Bio C4 columns.These Samplets are compatible with 50 to 120 gram columns and contain approximately 10 grams of C18 sorbent material. Shipped in packs of 20.
Catalog No. NC1607503 Supplier Biotage Supplier No. SAS04010100
Creating dried, adsorbed samples will improve the separation performance on any cartridge. Biotage Samplet are designed for quick and convenient loading of concentrated samples.After the sample has been applied to the Samplet, the solvent is allowed to evaporate. The Samplet is then inserted into the Biotage Sfär Duo columns and you are ready to go.Samplets also are used as guard columns for Biotage Sfar C18, Biotage Sfar Bio C18 and Biotage Sfar Bio C4 columns.These Samplets are compatible with 50 to 120 gram columns and contain approximately 10 grams of C18 sorbent material. Shipped in packs of 20.