Learn More
Biotage Sfär DLV Column with 10g of Silicia
SDP
Catalog No. NC1606645
Encompass
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. Users adsorb liquid samples onto the 10 grams of pre-packed adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20. These vessels come pre-packed with 10 grams of silica gel and have approximately 10 mL of headspace to accomdate liquid loading.Compatible with Biotage DLV plunger, part number DLV-025
Catalog No. NC1606645 Supplier Biotage Supplier No. DLV04450025
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. Users adsorb liquid samples onto the 10 grams of pre-packed adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20. These vessels come pre-packed with 10 grams of silica gel and have approximately 10 mL of headspace to accomdate liquid loading.Compatible with Biotage DLV plunger, part number DLV-025