Learn More
Biotage Sfär DLV Column with 5g of Silicia
SDP
Catalog No. NC1606636
Encompass
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. Users adsorb liquid samples onto the 5 grams of pre-packed adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20. These vessels come pre-packed with 5 grams of silica gel and have approximately 5 mL of headspace to accomdate liquid loading.Compatible with Biotage DLV plunger, part number DLV-010
Catalog No. NC1606636 Supplier Biotage Supplier No. DLV04450010
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. Users adsorb liquid samples onto the 5 grams of pre-packed adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20. These vessels come pre-packed with 5 grams of silica gel and have approximately 5 mL of headspace to accomdate liquid loading.Compatible with Biotage DLV plunger, part number DLV-010