Learn More
Biotage Sfär DLV Empty 10g Column with Frit
SDP
Catalog No. NC1606621
Encompass
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. With maximum flexibility, users place the needed amount of adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20 with 20 frits. These are empty vessels and can accomodate up to approximately 10 grams of most sorbants, such as silica gel.Compatible with Biotage part number DLV-010
Catalog No. NC1606621 Supplier Biotage Supplier No. DLV00000010
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. With maximum flexibility, users place the needed amount of adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20 with 20 frits. These are empty vessels and can accomodate up to approximately 10 grams of most sorbants, such as silica gel.Compatible with Biotage part number DLV-010