Learn More
Biotage Sfär DLV Empty 25g Column with Frit
SDP
Catalog No. NC1606622
Encompass
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. With maximum flexibility, users place the needed amount of adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20 with 20 frits. These are empty vessels and can accomodate up to approximately 25 grams of most sorbants, such as silica gel.Compatible with Biotage part number DLV-025
Catalog No. NC1606622 Supplier Biotage Supplier No. DLV00000025
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. With maximum flexibility, users place the needed amount of adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20 with 20 frits. These are empty vessels and can accomodate up to approximately 25 grams of most sorbants, such as silica gel.Compatible with Biotage part number DLV-025