Learn More
Biotage Sfär DLV 10g Column Plunger
SDP
Catalog No. NC1606629
Encompass
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. The 10 gram DLV plunger's innovative design attaches securely, compresses the sorbent bed and eliminates dead volume. Attachement to any Biotage Sfar column or flash chromatography column with luer fittings is secure. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Compatible with Biotage DLV vessels, part numbers DLV-0000-0010, DLV-0445-0010, and DLV-0825-0010.
Catalog No. NC1606629 Supplier Biotage Supplier No. DLV010
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. The 10 gram DLV plunger's innovative design attaches securely, compresses the sorbent bed and eliminates dead volume. Attachement to any Biotage Sfar column or flash chromatography column with luer fittings is secure. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Compatible with Biotage DLV vessels, part numbers DLV-0000-0010, DLV-0445-0010, and DLV-0825-0010.