Learn More
Biotage Sfär DLV 50g/100g Column Plunger
SDP
Catalog No. NC1606652
Encompass
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. The 50/100 gram DLV plunger's innovative design attaches securely, compresses the sorbent bed and eliminates dead volume. Attachement to any Biotage Sfar column or flash chromatography column with luer fittings is secure. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Compatible with Biotage DLV vessels, part numbers DLV-0000-0050, DLV-0445-0050, and DLV-0825-0050.
Catalog No. NC1606652 Supplier Biotage Supplier No. DLV050
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. The 50/100 gram DLV plunger's innovative design attaches securely, compresses the sorbent bed and eliminates dead volume. Attachement to any Biotage Sfar column or flash chromatography column with luer fittings is secure. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Compatible with Biotage DLV vessels, part numbers DLV-0000-0050, DLV-0445-0050, and DLV-0825-0050.