Learn More
Biotage Sfär Silica Samplet 50 to 120g Column
SDP
Catalog No. NC1607507
Encompass
Creating dried, adsorbed samples will improve the separation performance on any cartridge. Biotage Samplet are designed for quick and convenient loading of concentrated samples.After the sample has been applied to the Samplet, the solvent is allowed to evaporate. The Samplet is then inserted into the Biotage Sfär Duo columns and you are ready to go.These Samplets are compatible with 50 to 120 gram columns and contain approximately 10 grams of silica sorbent material. Shipped in packs of 20.
Catalog No. NC1607507 Supplier Biotage Supplier No. SAS04450100
Creating dried, adsorbed samples will improve the separation performance on any cartridge. Biotage Samplet are designed for quick and convenient loading of concentrated samples.After the sample has been applied to the Samplet, the solvent is allowed to evaporate. The Samplet is then inserted into the Biotage Sfär Duo columns and you are ready to go.These Samplets are compatible with 50 to 120 gram columns and contain approximately 10 grams of silica sorbent material. Shipped in packs of 20.