Learn More
Biotage Sfär Silica 60µm 5g
SDP
Catalog No. NC1606496
Encompass
Biotage Sfär Silica - 60 μm is the ideal solution for standard separations that require a reliable normal phase media and injection of samples in liquid form or using the Biotage DLV system.Sfär means “Sphere” in the Swedish mother tongue of Biotage and indicates that spherical silica is used in these columns. Spherical silica has many advantages, higher surface area, higher loading capacities, tighter elution bands and purer fractions. Sfär columns are quality tested to ensure they meet stringent performance criteria, including efficiency and peak symmetry. Each CE-marked column is created using inert, food grade plastics for lower extractables, cleaner fractions, and packed to provide excellent resolution. This is our sealed column which benefits from our highest pressure rating of 20 bar or 290 psi).These 5g columns come in convenient dispenser boxes containing 20 columns
Catalog No. NC1606496 Supplier Biotage Supplier No. FSRS04450005
Biotage Sfär Silica - 60 μm is the ideal solution for standard separations that require a reliable normal phase media and injection of samples in liquid form or using the Biotage DLV system.Sfär means “Sphere” in the Swedish mother tongue of Biotage and indicates that spherical silica is used in these columns. Spherical silica has many advantages, higher surface area, higher loading capacities, tighter elution bands and purer fractions. Sfär columns are quality tested to ensure they meet stringent performance criteria, including efficiency and peak symmetry. Each CE-marked column is created using inert, food grade plastics for lower extractables, cleaner fractions, and packed to provide excellent resolution. This is our sealed column which benefits from our highest pressure rating of 20 bar or 290 psi).These 5g columns come in convenient dispenser boxes containing 20 columns