Learn More
Biotage Sfär Silica Duo 60µm 10g
SDP
Catalog No. NC1606484
Encompass
Biotage Sfär Silica D - Duo 60 μm is the ideal solution for standard separations that require a reliable normal phase media and the flexibility of loading the samples in different ways.Sfär means “Sphere” in the Swedish mother tongue of Biotage and indicates that spherical silica is used in these columns. Spherical silica has many advantages, higher surface area, higher loading capacities, tighter elution bands and purer fractions. D stands for Duo, as Biotage Sfär gives you the dual possibility of liquid loading and drying loading, on the column.Sfär columns are quality tested to ensure they meet stringent performance criteria, including efficiency and peak symmetry. Each CE-marked column is created using inert, food grade plastics for lower extractables, cleaner fractions, and packed to provide excellent resolution alongside tolerance of higher operating pressures (as high as 17 bar or 247 psi).These 10g columns come in convenient dispenser boxes containing 20 columns
Catalog No. NC1606484 Supplier Biotage Supplier No. FSRD04450010
Biotage Sfär Silica D - Duo 60 μm is the ideal solution for standard separations that require a reliable normal phase media and the flexibility of loading the samples in different ways.Sfär means “Sphere” in the Swedish mother tongue of Biotage and indicates that spherical silica is used in these columns. Spherical silica has many advantages, higher surface area, higher loading capacities, tighter elution bands and purer fractions. D stands for Duo, as Biotage Sfär gives you the dual possibility of liquid loading and drying loading, on the column.Sfär columns are quality tested to ensure they meet stringent performance criteria, including efficiency and peak symmetry. Each CE-marked column is created using inert, food grade plastics for lower extractables, cleaner fractions, and packed to provide excellent resolution alongside tolerance of higher operating pressures (as high as 17 bar or 247 psi).These 10g columns come in convenient dispenser boxes containing 20 columns