Learn More
Biotage Sfär Silica HC 20µm 5g - High Capacity Column
SDP
Catalog No. NC1606560
Encompass
Biotage Sfär Silica High Capacity 20 μm is the choice when you need to run the best possible separation with the smallest column and greenest amount of solvent. This 5g high capacity column has the separation capacity of a 10g standard capacity column.Sfär means “Sphere” in the Swedish mother tongue of Biotage, and is the name for our columns as they contain spherical silica with higher surface area compared to irregular silica. Which means you use a smaller column with tight elution bands and pure fractions.Sfär columns are quality tested to ensure they meet stringent performance criteria, including efficiency and peak symmetry. Each CE-marked column is created using inert, food grade plastics for lower extractables, cleaner fractions, and packed to provide excellent resolution. This is our sealed column which benefits from our highest pressure rating of 20 bar or 290 psi.These 5g columns come in convenient dispenser boxes containing 20 columns
Catalog No. NC1606560 Supplier Biotage Supplier No. FSUS04430005
Biotage Sfär Silica High Capacity 20 μm is the choice when you need to run the best possible separation with the smallest column and greenest amount of solvent. This 5g high capacity column has the separation capacity of a 10g standard capacity column.Sfär means “Sphere” in the Swedish mother tongue of Biotage, and is the name for our columns as they contain spherical silica with higher surface area compared to irregular silica. Which means you use a smaller column with tight elution bands and pure fractions.Sfär columns are quality tested to ensure they meet stringent performance criteria, including efficiency and peak symmetry. Each CE-marked column is created using inert, food grade plastics for lower extractables, cleaner fractions, and packed to provide excellent resolution. This is our sealed column which benefits from our highest pressure rating of 20 bar or 290 psi.These 5g columns come in convenient dispenser boxes containing 20 columns