Learn More
New England Biolabs, Inc. SfcI – 1000 units
SDP
Catalog No. 50812246
Encompass
A restriction endonuclease that recognizes the sequence C^TRYA_G.
Catalog No. 50-812-246 Supplier New England Biolabs, Inc. Supplier No. R0561L
A restriction endonuclease that recognizes the sequence C^TRYA_G.