Learn More
New England Biolabs, Inc. SfcI – 200 units
SDP
Catalog No. 50812247
Encompass
A restriction endonuclease that recognizes the sequence C^TRYA_G.
Catalog No. 50-812-247 Supplier New England Biolabs, Inc. Supplier No. R0561S
A restriction endonuclease that recognizes the sequence C^TRYA_G.