Learn More
New England Biolabs, Inc. SfiI – 15000 units
SDP
Catalog No. 50811966
Encompass
A restriction endonuclease that recognizes the sequence GGCCN_NNN^NGGCC.
Catalog No. 50-811-966 Supplier New England Biolabs, Inc. Supplier No. R0123L
A restriction endonuclease that recognizes the sequence GGCCN_NNN^NGGCC.