Learn More
New England Biolabs, Inc. SfiI – 3000 units
SDP
Catalog No. 50811967
Encompass
A restriction endonuclease that recognizes the sequence GGCCN_NNN^NGGCC.
Catalog No. 50-811-967 Supplier New England Biolabs, Inc. Supplier No. R0123S
A restriction endonuclease that recognizes the sequence GGCCN_NNN^NGGCC.