Learn More
New England Biolabs, Inc. SfoI – 500 units
SDP
Catalog No. 50812334
Encompass
A restriction endonuclease that recognizes the sequence GGC_^GCC.
Catalog No. 50-812-334 Supplier New England Biolabs, Inc. Supplier No. R0606S
A restriction endonuclease that recognizes the sequence GGC_^GCC.