Learn More
Aviva Systems Biology Corp SFPQ Antibody - middle region (ARP40571_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40571_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SFPQ. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SFPQ. Alternate names: POMP100, PSFApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57527