Learn More
Aviva Systems Biology Corp SFPQ Antibody - N-terminal region (ARP40572_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40572_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SFPQ. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SFPQ. Alternate names: PSF, POMP100ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-57528