Learn More
Aviva Systems Biology Corp SFRP1 Antibody - middle region (AVARP09053_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09053_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SFRP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SFRP1. Alternate names: FRP, FRP1, FrzA, FRP-1, SARP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53777