Learn More
Medchemexpress LLC SFRP1 Mouse CHO H 50ug
SDP
Catalog No. 5000180359
Encompass_Preferred
SFRP1 Protein is a secreted glycoprotein member of the SFRP family SFRP1 Protein can act as an inhibitor of Wnt signaling and is capable of resisting vascular cell proliferation SFRP1 Protein Mouse (CHO His) is the recombinant mouse-derived SFRP1 protein expressed by CHO with C-10 His labeled tag
Catalog No. 50-001-80359 Supplier Medchemexpress LLC Supplier No. HYP7341350UG
SFRP1 Protein is a secreted glycoprotein member of the SFRP family SFRP1 Protein can act as an inhibitor of Wnt signaling and is capable of resisting vascular cell proliferation SFRP1 Protein Mouse (CHO His) is the recombinant mouse-derived SFRP1 protein expressed by CHO with C-10 His labeled tag