Learn More
Sino Biological Recombinant Mouse SFRP1 Protein (His Tag)
Supplier:  Sino Biological 50033M08S50
Encompass_Preferred
A DNA sequence encoding the mouse SFRP1 (AAC53145.1) (Met1-Lys314) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4796