Learn More
Aviva Systems Biology Corp SFRS1 Antibody - C-terminal region (ARP40691_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40691_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFRS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SFRS1. Alternate names: ASF, SF2, SFRS1, SF2p33, SRp30a, SRSF1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57589