Learn More
Aviva Systems Biology Corp SFRS10 Antibody - middle region (ARP40529_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40529_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SFRS10. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SFRS10. Alternate names: SFRS10, SRFS10, TRAN2B, TRA2-BETA, Htra2-betaApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57506