Learn More
Aviva Systems Biology Corp SFRS10 Antibody - N-terminal region (ARP40528_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40528_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SFRS10. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SFRS10. Alternate names: SFRS10, SRFS10, TRAN2B, TRA2-BETA, Htra2-betaApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57505