Learn More
Aviva Systems Biology Corp SFRS12IP1 Antibody - middle region (ARP55719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFRS12IP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SFRS12IP1. Alternate names: FLJ36754, MGC131910, MGC150548, MGC150549, P18SRP, SFRS12IP1ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-61589