Learn More
Aviva Systems Biology Corp SFRS12IP1 Antibody - N-terminal region (ARP55718_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55718_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFRS12IP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SFRS12IP1. Alternate names: FLJ36754, MGC131910, MGC150548, MGC150549, P18SRP, SFRS12IP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-286-070