Learn More
Aviva Systems Biology Corp SFRS4 Antibody - middle region (ARP40591_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40591_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFRS4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SFRS4. Alternate names: SRP75, SFRS4ApplicationWB. Species reactivity: Dog, Human, Pig, Rat
Catalog No. 50-014-57538