Learn More
ABclonal Technology SFT Rabbit mAb
SDP
Catalog No. 5000008007
Encompass_Preferred
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation Ubiquitination involves at least three classes of enzymes ubiquitin-activating enzymes or E1s ubiquitin-conjugating enzymes or E2s and ubiquitin-protein ligases or E3s This gene encodes a member of the E2 ubiquitin-conjugating enzyme family This enzyme is closely related to a stimulator of iron transport (SFT) and is up-regulated in hereditary hemochromatosis It also functions in the ubiquitination of the tumor-suppressor protein p53 and the hypoxia-inducible transcription factor HIF1alpha by interacting with the E1 ubiquitin-activating enzyme and the E3 ubiquitin-protein ligases Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-08007 Supplier ABclonal Technology Supplier No. A21076100UL
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation Ubiquitination involves at least three classes of enzymes ubiquitin-activating enzymes or E1s ubiquitin-conjugating enzymes or E2s and ubiquitin-protein ligases or E3s This gene encodes a member of the E2 ubiquitin-conjugating enzyme family This enzyme is closely related to a stimulator of iron transport (SFT) and is up-regulated in hereditary hemochromatosis It also functions in the ubiquitination of the tumor-suppressor protein p53 and the hypoxia-inducible transcription factor HIF1alpha by interacting with the E1 ubiquitin-activating enzyme and the E3 ubiquitin-protein ligases Two transcript variants encoding different isoforms have been found for this gene