Learn More
MSC Shepherd Regent Stem Mount Swivel Caster, Rubber, 3 in. Dia, 150 lb. Capacity, 1 ea
Catalog No. 5000514255
Encompass
This stem mount swivel caster is designed for industrial and institutional applications. Featuring a rubber wheel construction and a zinc finish, it provides reliable mobility for various transport needs.
  • Wheel diameter of 3 in.
  • Width of 1-1/4 in.
  • Overall height of 4-1/8 in.
  • Load capacity of 150 lb.
  • Equipped with a 1/2-13 x 1-1/2 in. threaded stem
  • Includes nylon bearing for smooth swivel operation
Catalog No. 50-005-14255 Supplier MSC Supplier No. 79603015
This stem mount swivel caster is designed for industrial and institutional applications. Featuring a rubber wheel construction and a zinc finish, it provides reliable mobility for various transport needs.
  • Wheel diameter of 3 in.
  • Width of 1-1/4 in.
  • Overall height of 4-1/8 in.
  • Load capacity of 150 lb.
  • Equipped with a 1/2-13 x 1-1/2 in. threaded stem
  • Includes nylon bearing for smooth swivel operation