Learn More
Aviva Systems Biology Corp SFT2D3 Antibody - N-terminal region (ARP60656_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60656_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFT2D3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SFT2D3. Alternate names: MGC5391ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-287-117