Learn More
Aviva Systems Biology Corp SFTPB Antibody - middle region (ARP41411_P050) | 100uL
SDP
Catalog No. 5001457839
Encompass_Preferred
This is a rabbit polyclonal antibody against SFTPB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SFTPB. Alternate names: PSP-B, SFTB3, SFTP3, SMDP1, SP-BApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-57839 Supplier Aviva Systems Biology Corp Supplier No. ARP41411_P050
This is a rabbit polyclonal antibody against SFTPB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SFTPB. Alternate names: PSP-B, SFTB3, SFTP3, SMDP1, SP-BApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep