Learn More
Aviva Systems Biology Corp SFTPD Antibody - middle region (ARP41514_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41514_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFTPD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SFTPD. Alternate names: COLEC7, PSP-D, SFTP4, SP-DApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57890