Learn More
ACROBiosystems SFTS virus Gn protein 50ug
Catalog No. 502534956
Encompass_Preferred
SFTS virus Gn Protein (HN6) His Tag (GNN-S52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Lys 452 (Accession # F1BWU4).
Catalog No. 50-253-4956 Supplier ACROBiosystems Supplier No. GNNS52H450UG
SFTS virus Gn Protein (HN6) His Tag (GNN-S52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Lys 452 (Accession # F1BWU4).