Learn More
MSC MCR Safety Glasses, Over-the-Glasses, Anti-Fog Clear, ANSI Z87.1, 1 Pair
Catalog No. 501909738
Encompass
These safety glasses are designed to fit comfortably over most prescription eyewear, offering robust protection with clear, anti-fog polycarbonate lenses. They are suitable for various industrial and laboratory environments where eye protection is required.
  • Oversized frame and lens for compatibility with prescription glasses
  • Dielectric construction with no metal components
  • Ratchet action temple with 5 positions for adjustable lens angle
  • Duramass® scratch-resistant coating enhances durability
  • Polycarbonate lens filters 99.9% of harmful UV radiation
  • Anti-fog coatings maintain clear vision in challenging conditions
  • Meets ANSI Z87.1 safety standards
This item is not returnable. View return policy
Catalog No. 50-190-9738 Supplier MSC Supplier No. 68872225
These safety glasses are designed to fit comfortably over most prescription eyewear, offering robust protection with clear, anti-fog polycarbonate lenses. They are suitable for various industrial and laboratory environments where eye protection is required.
  • Oversized frame and lens for compatibility with prescription glasses
  • Dielectric construction with no metal components
  • Ratchet action temple with 5 positions for adjustable lens angle
  • Duramass® scratch-resistant coating enhances durability
  • Polycarbonate lens filters 99.9% of harmful UV radiation
  • Anti-fog coatings maintain clear vision in challenging conditions
  • Meets ANSI Z87.1 safety standards