Learn More
Medicus Health FLEX ARM SAFETY SPLASH SHIELD, 24" WHITE GOOSENECK ARM
SDP
Catalog No. NC1426593
Encompass
∙ Flex Arm Safety Splash Shield with 18" Gooseneck Arm and C-clamp mounting base allows for precise positioning∙Flat Splash Shield mounted on a white vinyl coated flexible arm ∙Ultimate in flexibility for optimum protection with a 10" x 10" flat shield∙18" White Gooseneck arm allows for precise positioning. ∙ 10” x 10” flat shield is mounted diagonally so it adds another 13” to the overall length of the shield.∙Coated arm is easy to clean and keep clean∙Durable, scratch-resistant polycarbonate shield protects you from splashes, aerosol mist, and flying debris∙Helps meet compliance with OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)∙C-clamp mount is padded to protect your desk or bench top from scratches and can be mounted on a surface up to 2" thick∙To create a permanent mount, simply remove the C-clamp and fasten to any solid surface
This item is not returnable. View return policy
Catalog No. NC1426593 Supplier Medicus Health Supplier No. 5270M3
∙ Flex Arm Safety Splash Shield with 18" Gooseneck Arm and C-clamp mounting base allows for precise positioning∙Flat Splash Shield mounted on a white vinyl coated flexible arm ∙Ultimate in flexibility for optimum protection with a 10" x 10" flat shield∙18" White Gooseneck arm allows for precise positioning. ∙ 10” x 10” flat shield is mounted diagonally so it adds another 13” to the overall length of the shield.∙Coated arm is easy to clean and keep clean∙Durable, scratch-resistant polycarbonate shield protects you from splashes, aerosol mist, and flying debris∙Helps meet compliance with OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)∙C-clamp mount is padded to protect your desk or bench top from scratches and can be mounted on a surface up to 2" thick∙To create a permanent mount, simply remove the C-clamp and fasten to any solid surface