Learn More
Med Vet International Exel Safety Scalp Vein Set, with 21g x 3/4in Needle, 12in Tubing, 50/box
Supplier:  Med Vet International 27704
Encompass_Preferred
Exel International's safety scalp vein set has a 21g x 3/4in. thin wall stainless steel cannula, that is siliconized for easy insertion. It has a 12in. tube with flexible wings that hold the needle in position and helps to prevent inadvertent pull out. This infusion set has a green-colored hub for easy identification. Each scalp vein set in this box is latex free and individually packaged.
Catalog No. 50-255-9724