Learn More
Aviva Systems Biology Corp SFXN1 Antibody - N-terminal region (ARP49750_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49750_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFXN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SFXN1. Alternate names: FLJ12876ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-60167