Learn More
Aviva Systems Biology Corp SFXN4 Antibody - N-terminal region (ARP53530_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53530_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFXN4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SFXN4. Alternate names: BCRM1ApplicationWB. Species reactivity: Human
Catalog No. 50-285-846