Learn More
Lonza Walkersville SG Cell Line 384-well Nucleofector Kit
Catalog No. NC2284651
Encompass
The SG Cell Line 384-well Nucleofector Kit is one of our three kits suited for high-throughput transfection of cell lines when working with the 384-well HT Nucleofector System.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCells maintain functionality post transfectionSeamless integration into automated liquid handling environmentsConditions are transferable between 4D-Nucleofector System, 96-well Shuttle System and 384-well Nucleofector SystemApplications:Transfection of low cell numbers down to 2 x 10⁴ cell linesHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:90 mL SG Cell Line Nucleofector Solution20 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)10 x 384-well Nucleocuvette Plate (20 μl)
This item is not returnable. View return policy
Catalog No. NC2284651 Supplier Lonza Walkersville Supplier No. V5SC3010
Add to Cart
Add to Cart
Request a Quote
The SG Cell Line 384-well Nucleofector Kit is one of our three kits suited for high-throughput transfection of cell lines when working with the 384-well HT Nucleofector System.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCells maintain functionality post transfectionSeamless integration into automated liquid handling environmentsConditions are transferable between 4D-Nucleofector System, 96-well Shuttle System and 384-well Nucleofector SystemApplications:Transfection of low cell numbers down to 2 x 10⁴ cell linesHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:90 mL SG Cell Line Nucleofector Solution20 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)10 x 384-well Nucleocuvette Plate (20 μl)