Learn More
Molecular Dimensions SG1 (SHOTGUN) SCREEN HT-96
SDP
Catalog No. NC0791270
Encompass
Protein Crystallography. SG1 (SHOTGUN) SCREEN HT-96 (96 x 1.1mL): 96 conditions that occur with highest, non-redundant frequency amongst all PDB deposits. SG1 uses the ShotGun approach to gather all the most successful conditions from all the early commercially available screens.
Catalog No. NC0791270 Supplier Molecular Dimensions Supplier No. MD189
Protein Crystallography. SG1 (SHOTGUN) SCREEN HT-96 (96 x 1.1mL): 96 conditions that occur with highest, non-redundant frequency amongst all PDB deposits. SG1 uses the ShotGun approach to gather all the most successful conditions from all the early commercially available screens.