Learn More
Medchemexpress LLC SG3199 | 1595275-71-0 | MFCD34469130 | >98.0% | 584.66 g/mol | C33H36N4O6 | 10 MG
SDP
Catalog No. 5000212192
Encompass_Preferred
SG3199 is a cytotoxic pyrrolobenzodiazepine (PBD) dimer that induces DNA minor-groove interstrand cross-links. It serves as a potent warhead for antibody-drug conjugate research and shows picomolar cytotoxicity in cancer cell lines. Handle with appropriate laboratory safety and storage conditions.
  • Potent cytotoxicity in cancer cell lines (mean GI50 ≈ 151.5 pM).
  • DNA minor-groove interstrand cross-linking mechanism.
  • High DMSO solubility for in vitro assay preparation.
  • Requires protection from light and storage under nitrogen at -20°C; in solvent, -80°C for long-term storage.
  • Suitable for ADC payload and mechanism-of-action studies.
Catalog No. 50-002-12192 Supplier Medchemexpress LLC Supplier No. HY10116110MG
SG3199 is a cytotoxic pyrrolobenzodiazepine (PBD) dimer that induces DNA minor-groove interstrand cross-links. It serves as a potent warhead for antibody-drug conjugate research and shows picomolar cytotoxicity in cancer cell lines. Handle with appropriate laboratory safety and storage conditions.
  • Potent cytotoxicity in cancer cell lines (mean GI50 ≈ 151.5 pM).
  • DNA minor-groove interstrand cross-linking mechanism.
  • High DMSO solubility for in vitro assay preparation.
  • Requires protection from light and storage under nitrogen at -20°C; in solvent, -80°C for long-term storage.
  • Suitable for ADC payload and mechanism-of-action studies.