Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SG3199-Val-Ala-PAB | 1595275-61-8 | 96.9% | 922.03 g/mol | C49H59N7O11 | 25 MG
SDP

Catalog No. 5000213143
Encompass_Preferred

SG3199-Val-Ala-PAB is a synthetic intermediate used in the synthesis of Tesirine, an agent-linker conjugate employed in antibody-drug conjugate (ADC) research. The material is provided as a high-purity research reagent for linker-payload development and preclinical studies.

  • Intermediate for ADC linker-payload synthesis
  • High purity (96.9%) suitable for research use
  • Molecular weight 922.03 g/mol for formulation calculations
  • CAS number available for unambiguous identification
  • Offered in milligram-scale pack sizes for discovery work
  • Soluble in DMSO for convenient stock solution preparation

Catalog No. 50-002-13143 Supplier Medchemexpress LLC Supplier No. HY10115225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SG3199-Val-Ala-PAB is a synthetic intermediate used in the synthesis of Tesirine, an agent-linker conjugate employed in antibody-drug conjugate (ADC) research. The material is provided as a high-purity research reagent for linker-payload development and preclinical studies.

  • Intermediate for ADC linker-payload synthesis
  • High purity (96.9%) suitable for research use
  • Molecular weight 922.03 g/mol for formulation calculations
  • CAS number available for unambiguous identification
  • Offered in milligram-scale pack sizes for discovery work
  • Soluble in DMSO for convenient stock solution preparation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.