Learn More
Apexbio Technology LLC SGC 0946, 10mg. Cas: 1561178-17-3 MFCD: MFCD26523136. DOT1L inhibitor,highly potent and selective
SDP
Supplier:  Apexbio Technology LLC A416710
Encompass_Preferred
SGC 0946 is a potent small-molecule inhibitor of protein methyltransferase (PMT) DOT1L, a unique protein lysine methyltransferase (PKMT) catalyzing the methylation of histone H3 on Lysine 79 (H3K79), with the half maximal inhibition concentration IC50 value of 0.3 nM [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4898
 Restricted Item