Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGC 0946, Tocris Bioscience™

Catalog No. 454150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-415-0 50 mg
45-411-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-415-0 Supplier R&D Systems Supplier No. 4541/50
This item is not returnable. View return policy

Highly potent and selective DOT1L inhibitor; cell permeable

Highly potent DOT1L methyltransferase inhibitor (Kd = 0.06 nM, IC50 = 0.3 nM in a radioactive assay); blocks H3K79 methylation in A431 cells and MCF10A cells. Inactive at 12 histone methyltransferases and DNMT1. Selectively kills cells transformed with the MLL-AF9 fusion oncogene in an in vitro model of leukemia; lowers levels of MLL target genes HOXA9 and Meis1.

To request the negative control for SGC0946, please fill out the SGC0649 request form on the SGC website.

Chemical Identifiers

CAS 1561178-17-3
Molecular Formula C28H40BrN7O4
Molecular Weight (g/mol) 618.58
InChI Key IQCKJUKAQJINMK-UHFFFAOYNA-N
Synonym 1-3-2r,3s,4r,5r-5-4-amino-5-bromo-7h-pyrrolo 2,3-d pyrimidin-7-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl isopropyl amino propyl-3-4-tert-butyl phenyl urea, d0s9sj, sgc hplc, 1-3-2r,3s,4r,5r-5-4-amino-5-bromopyrrolo 2,3-d pyrimidin-7-yl-3,4-dihydroxyoxolan-2-yl methyl-propan-2-ylamino propyl-3-4-tert-butylphenyl urea, 1-3-2r,3s,4r,5r-5-4-amino-5-bromo-7h-pyrrolo 2,3-d pyrimidin-7-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl isopropyl amino propyl-3-4-2,2-dimethylethyl phenyl urea, 5-bromo-7-5-deoxy-5-3-4-1,1-dimethylethyl phenyl amino carbonyl amino propyl 1-methylethyl amino-beta-d-7h-pyrrolo 2,3-d pyrimidin-4-amine, 5-bromo-7-5-3-4-tert-butylphenyl carbamoyl amino propyl propan-2-yl amino-5-deoxy-beta-d-ribofuranosyl-7h-pyrrolo 2,3-d pyrimidin-4-amine, aw2
PubChem CID 56962337

Specifications

Quantity 50 mg
Formula Weight 618.57
Percent Purity >99%
Chemical Name or Material SGC 0946
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.