Learn More
Enzo Life Sciences Guanylyl cyclase alpha1beta1 (soluble) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210786R100
Encompass_Preferred
Host: Chicken. Immunogen: Purified rat lung soluble guanylyl cyclase α1β1 (sGCα1β1) (native protein). Species reactivity: Human, Mouse, Rat. Specificity: Recognizes endogenous rat sGCα1β1. Recognizes human and mouse sGCβ1. Applications: WB. Purity: Purified IgY from egg yolk. Formulation: Liquid. In 10mM TRIS-HCl, pH 7.5, containing 150mM sodium chloride and 0.02% thimerosal.
Catalog No. 50-200-8154