Learn More
Selleck Chemical LLC SGC-CBP30 10mg 1613695-14-9
SDP
Supplier:  Selleck Chemical LLC S725610MG
Encompass_Preferred
SGC-CBP30 is a potent CREBBP/EP300 inhibitor with IC50 of 21 nM and 38 nM in cell-free assays, respectively. Exhibits 40-fold and 250-fold selectivity for CBP over the first BRD of BRD4 (BRD4(1)) and BRD4(2) respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4560